Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc protoporphyrin IX, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

15442-64-5
C34H32N4O4Zn
626.034
FUTVBRXUIKZACV-UHFFFAOYSA-L
7,12-Diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropionic acid, zinc complex
131664383

Heme oxygenase and guanylyl cyclase inhibitor

Inhibitor of heme oxygenase, which generates the putative neurotransmitter CO.
SMILES CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2]
IUPAC Name zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate
Formula Weight 626.03
Quantity 50mg
Chemical Name or Material Zinc protoporphyrin IX
Grade >98%

Recommended Storage : Store at Room Temperature