Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc protoporphyrin IX, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

15442-64-5
C34H32N4O4Zn
626.034
FUTVBRXUIKZACV-UHFFFAOYSA-L
7,12-Diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropionic acid, zinc complex
131664383
zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2]

Heme oxygenase and guanylyl cyclase inhibitor

Inhibitor of heme oxygenase, which generates the putative neurotransmitter CO.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 626.03
Quantity 10mg
Chemical Name or Material Zinc protoporphyrin IX
Grade >98%

Recommended Storage : Store at Room Temperature