Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Pyrithione, 48% Aqueous Suspension, Spectrum™ Chemical
SDP

Catalog Number 18607871
Click to view available options
Quantity:
100 g
500 g
2.5 kg
5 kg
12 kg
Packaging:
Amber Glass Bottle
Glass Bottle
Poly Pail
13463-41-7
C10H8N2O2S2Zn
317.69
OTPSWLRZXRHDNX-UHFFFAOYSA-L
zinc pyrithione
[Zn++].[O-][N+]1=CC=CC=C1[S-].[O-][N+]1=CC=CC=C1[S-]
13463-41-7
C10H8N2O2S2Zn
317.69
OTPSWLRZXRHDNX-UHFFFAOYSA-L
zinc pyrithione
[Zn++].[O-][N+]1=CC=CC=C1[S-].[O-][N+]1=CC=CC=C1[S-]

Z1025, 13463-41-7

Spectrum™ Chemical Zinc Pyrithione, 48 Percent Aqueous Suspension is a fungistatic and baceriostatic compound that has a variety of uses. It is used in outdoor paint and other products that provide protection against mildew and algae due to its low solubility in water (8 ppm at neutral pH). It is also often used in kitchen sponges as an antibacterial treatment. 48 Percent Aqueous Suspension.

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 48 to 52%
Grade Reagent
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit