Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc selenide, Optical Grade, 99.999% (metals basis)

For vacuum deposition

Manufacturer:  Alfa Aesar™ 8829814

Quantity 25g

 View more versions of this product

Catalog No. AA8829814


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1315-09-9
144.33
UN3283
Zinc selenide
ZnSe
MFCD00011296
14,10156
Optical Grade

Specifications

1315-09-9
ZnSe
MFCD00011296
14,10156
Odorless
Air Sensitive
25g
99.999%, (metals basis)
Pieces
144.33
UN3283
5.42g/mL at 15°C
3 to 6mm
>1100°C
Zinc selenide
Optical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P273-P270-P309-P310-P302+P352-P501a
Avoid release to the environment.
Do not eat, drink or smoke when using this product.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit