Alfa Aesar™ Zinc selenide, Optical Grade, 99.999% (metals basis)

For vacuum deposition

Manufacturer: Alfa Aesar™ 8829806

Quantity 5g

 View more versions of this product

Catalog No. AA8829806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1315-09-9
    Molecular Formula ZnSe
    Formula Weight 144.33
    MDL Number MFCD00011296
    UN Number UN3283
    Merck Index 14,10156
    Chemical Name or Material Zinc selenide
    Grade Optical Grade

    Specifications

    Formula Weight 144.33
    Health Hazard 3 GHS P Statement
    P273-P270-P309-P310-P302+P352-P501a
    Avoid release to the environment.
    Do not eat, drink or smoke when using this product.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011296
    CAS 1315-09-9
    Health Hazard 1 Danger
    TSCA TSCA
    Sensitivity Air Sensitive
    Molecular Formula ZnSe
    Melting Point >1100°C
    Quantity 5g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
    Odor Odorless
    UN Number UN3283
    Merck Index 14,10156
    Density 5.42g/mL at 15°C
    Particle Size 3 to 6mm
    Physical Form Pieces
    Health Hazard 2 GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Zinc selenide
    Assay 99.999%, (metals basis)
    Grade Optical Grade

    GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P273-P270-P309-P310-P302+P352-P501a
    Avoid release to the environment.
    Do not eat, drink or smoke when using this product.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit