Alfa Aesar™ Zinc selenite

Manufacturer: Alfa Aesar™ 1258609

Quantity 10g

 View more versions of this product

Catalog No. AA1258609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13597-46-1
    Formula Weight 192.33
    MDL Number MFCD00011297
    UN Number UN2630
    Chemical Name or Material Zinc selenite

    Specifications

    Formula Weight 192.33
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011297
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
    TSCA TSCA
    Physical Form Powder
    CAS 13597-46-1
    UN Number UN2630
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Zinc selenite

    GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit