Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Standard, 1mL = 1mg Zn, 1000ppm Zn, Ricca Chemical

Click to view available options
Quantity:
120mL
500mL
Packaging:
Poly natural
7440-66-6,7697-37-2,7732-18-5
Zn
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
CHEBI:30185
zinc
[Zn]
7440-66-6,7697-37-2,7732-18-5
Zn
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
CHEBI:30185
zinc
[Zn]
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 1,000ppm Zn
Equivalence 1 mL = 1 mg Zn
Color Colorless
Quantity 500mL
Chemical Name or Material Zinc Standard