Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Stearate, Powder, USP, 12.5-14%, Spectrum™

Click to view available options
Quantity:
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
557-05-1
C36H70O4Zn
632.33
XOOUIPVCVHRTMJ-UHFFFAOYSA-L
zinc(2+) dioctadecanoate
[Zn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
557-05-1
C36H70O4Zn
632.33
XOOUIPVCVHRTMJ-UHFFFAOYSA-L
zinc(2+) dioctadecanoate
[Zn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

ZI115, 557-05-1

Spectrum™ Zinc Stearate, Powder, USP is a mild antiseptic and astringent used as a local soothing application for inflammatory and irritating skin diseases and in pharmacuetical formulations as a lubricant in tablet and capsule manufacture. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Percent Purity 12.5 to 14%
Lead (Pb) 10 ppm
Packaging Fiber Drum
Quantity 45 kg