Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate, 7-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7446-20-0
H14O11SZn
287.541
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
CHEBI:32312

High quality chemicals for laboratory and specialized industrial use

SMILES O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
IUPAC Name zinc;sulfate;heptahydrate
Formula Weight 287.56
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.97kg
Quantity 125g
Chemical Name or Material Zinc Sulfate, 7-Hydrate
Grade Baker Analyzed™ ACS Reagent