Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate, Monohydrate, FCC, 98-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7446-19-7
H2O5SZn
179.45
RNZCSKGULNFAMC-UHFFFAOYSA-L
zinc(2+) hydrate sulfate
O.[Zn++].[O-]S([O-])(=O)=O
7446-19-7
H2O5SZn
179.45
RNZCSKGULNFAMC-UHFFFAOYSA-L
zinc(2+) hydrate sulfate
O.[Zn++].[O-]S([O-])(=O)=O

Z1100, 7446-19-7, ZnSO4•H2O

Spectrum™ Zinc Sulfate, Monohydrate, FCC is used as a nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Percent Purity 98.0 to 100.5%
Cadmium (Cd) 5 mg/kg
Lead (Pb) 4 mg/kg
Mercury (Hg) 5 mg/kg
Packaging Poly Pail
Selenium (Se) 3e-005
Quantity 12 kg