Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc sulfate monohydrate, Zn 35.5%

Manufacturer:  Alfa Aesar™ 14160A1

Quantity 1kg

 View more versions of this product

Catalog No. AA14160A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7446-19-7
MFCD00149893
14,10159
179.45 (161.43 Anhydrous)
UN3077
Zinc sulfate monohydrate

Specifications

7446-19-7
179.45 (161.43 Anhydrous)
UN3077
Soluble in water
1kg
Coarse Powder
MFCD00149893
14,10159
Zn 35.5%
Zinc sulfate monohydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280i-P273-P301+P310-P305+P351+P338-P315-P501a
Wear eye/face protection.
Avoid release to the environment.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Get immediate medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit