Alfa Aesar™ Zinc tungsten oxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4023430

Quantity 250g

 View more versions of this product

Catalog No. AA4023430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13597-56-3
    Formula Weight 313.22
    MDL Number MFCD00049633
    Synonym Zinc tungstate
    Chemical Name or Material Zinc tungsten oxide

    Specifications

    Formula Weight 313.22
    Health Hazard 3 GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H335
    May cause respiratory irritation.
    Mesh Size −200 Mesh
    Physical Form Powder
    MDL Number MFCD00049633
    CAS 13597-56-3
    Synonym Zinc tungstate
    Quantity 250g
    Health Hazard 1 Warning
    TSCA TSCA
    Chemical Name or Material Zinc tungsten oxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H335
    May cause respiratory irritation.

    GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit