Alfa Aesar™ Zinc wire, 1.0mm (0.04 in.) dia., Puratronic™, 99.9985% (metals basis)

Manufacturer: Alfa Aesar™ 12053CP

Quantity 2 x 500cm

 View more versions of this product

Catalog No. AA12053CP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-66-6
    MDL Number MFCD00011291
    Chemical Name or Material Zinc wire, 1.0mm (0.04 in.) dia.
    Grade Puratronic™

    Specifications

    TSCA TSCA
    CAS 7440-66-6
    Quantity 2 x 500cm
    Physical Form Wire
    MDL Number MFCD00011291
    Odor Odorless
    Weight ≈5.60g/m
    Chemical Name or Material Zinc wire, 1.0mm (0.04 in.) dia.
    Assay 99.9985%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit