Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ziprasidone hydrochloride, Tocris Bioscience™

Catalog Number p-4784856
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
122883-93-6
C21H24Cl2N4O2S
467.409
ZCBZSCBNOOIHFP-UHFFFAOYSA-N
ziprasidone hydrochloride monohydrate, ziprasidone hydrochloride, geodon, 5-2-4-benzo d isothiazol-3-yl piperazin-1-yl ethyl-6-chloroindolin-2-one hydrochloride hydrate, unii-216x081oru, ziprasidone hcl hydrate, ziprasidone hydrochloride usan:usp, zeldox, ziprasidone hydrochloride usan
60853
CHEBI:32314
5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one;hydrate;hydrochloride
C1CN(CCN1CCC2=C(C=C3C(=C2)CC(=O)N3)Cl)C4=NSC5=CC=CC=C54.O.Cl
This item is not returnable. View return policy
122883-93-6
C21H24Cl2N4O2S
467.409
ZCBZSCBNOOIHFP-UHFFFAOYSA-N
ziprasidone hydrochloride monohydrate, ziprasidone hydrochloride, geodon, 5-2-4-benzo d isothiazol-3-yl piperazin-1-yl ethyl-6-chloroindolin-2-one hydrochloride hydrate, unii-216x081oru, ziprasidone hcl hydrate, ziprasidone hydrochloride usan:usp, zeldox, ziprasidone hydrochloride usan
60853
CHEBI:32314
5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one;hydrate;hydrochloride
C1CN(CCN1CCC2=C(C=C3C(=C2)CC(=O)N3)Cl)C4=NSC5=CC=CC=C54.O.Cl

5-HT2A/D2 antagonist; atypical antipsychotic

Atypical antipsychotic that displays combined serotonin and dopamine receptor antagonism. Displays high affinity at 5-HT2A receptors with a 5-HT2A/D2 affinity ratio greater than any other clinically available atypical antipsychotics (pKi values are 9.38, 8.88, 8.69, 8.47, 8.32, 8.14, 7.98, 7.49, 7.33 and 6.28 for 5-HT2A, 5-HT2C, 5-HT1D, 5-HT1A, D2, D3, α1, D4, H1 and D1 receptors respectively).
Quantity 10 mg
Formula Weight 449.4
Percent Purity >99%
Chemical Name or Material Ziprasidone hydrochloride
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit