Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium Crucible

Manufacturer:  Alfa Aesar™ 35903KT

 View more versions of this product

Catalog No. AA35903KT


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Top: 39mm; Bottom: 29mm
With SnapOn Ring;Rign/ Rim OD: 44mm
Zirconium Crucible
33mm
Zirconium
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit