Alfa Aesar™ Zirconium crystal bar turnings, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 4722614

Quantity 25g

 View more versions of this product

Catalog No. AA4722614

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-67-7
    MDL Number MFCD00011303
    Chemical Name or Material Zirconium crystal bar turnings

    Specifications

    TSCA TSCA
    Odor Odorless
    Quantity 25g
    MDL Number MFCD00011303
    CAS 7440-67-7
    Chemical Name or Material Zirconium crystal bar turnings
    Assay 99.5%, (metals basis)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit