Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium Dichloride Oxide Hydrate, 99.9985% (Metal Basis), Thermo Scientific™, Puratronic™

Catalog Number p-4782245
Click to view available options
Quantity:
10 g
50 g
250 g
15461-27-5
Cl2OZr
178.12
MFCD00149898
IPCAPQRVQMIMAN-UHFFFAOYSA-L
zirconyl chloride hydrate, zirconium dichloride oxide hydrate, zirconium, dichlorooxo-, monohydrate, zirconium oxychloride hydrate, cl2ozr.h2o, zirconium dichloride oxide hydrate, puratronic, zirconyl chloride hydrate trace metals basis
16211499
Cl[Zr](Cl)=O
15461-27-5
Cl2OZr
178.12
MFCD00149898
IPCAPQRVQMIMAN-UHFFFAOYSA-L
zirconyl chloride hydrate, zirconium dichloride oxide hydrate, zirconium, dichlorooxo-, monohydrate, zirconium oxychloride hydrate, cl2ozr.h2o, zirconium dichloride oxide hydrate, puratronic, zirconyl chloride hydrate trace metals basis
16211499
Cl[Zr](Cl)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.9985%, metal basis
Solubility Information Soluble in cold water,methanol,diethyl ether and alcohol.Slightly soluble in hydrochloric acid. Insoluble in organic solvents.
IUPAC Name oxozirconium;hydrate;dihydrochloride
Formula Weight 178.13 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Zirconium Dichloride Oxide Hydrate
Hazard Category H302-H314-H335
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit