Learn More
Zirconium dinitrate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Zirconium dinitrate oxide hydrate is used as a precursor in the production of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and silicaand alumina-doped nanocrystalline ceria/zirconia. It is also used to prepare cerium oxide/zirconium oxide mixed oxide catalyst supports.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Soluble in alcohol. |
IUPAC Name | oxozirconium(2+);dinitrate;hydrate |
Formula Weight | 231.23 (Anhydrous) |
Chemical Name or Material | Zirconium dinitrate oxide hydrate |
Safety and Handling
Hazard Category | H272-H314 |
Precautionary Statement | P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 5.1; Packaging Group: III |
EINECSNumber | 237-529-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.