Alfa Aesar™ Zirconium dinitrate oxide hydrate, Puratronic™, 99.994% (metals basis)

Manufacturer: Alfa Aesar™ 1297106

Quantity 5g

 View more versions of this product

Catalog No. AA1297106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14985-18-3
    Linear Formula ZrO(NO3)2·xH2O
    Formula Weight 231.23 (Anhydrous)
    MDL Number MFCD00149896
    UN Number UN1477
    Chemical Name or Material Zirconium dinitrate oxide hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 231.23 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00149896
    UN Number UN1477
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H315-H319
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
    Odor Odorless
    Physical Form Powder
    Linear Formula ZrO(NO3)2·xH2O
    Color White
    Quantity 5g
    CAS 14985-18-3
    Chemical Name or Material Zirconium dinitrate oxide hydrate
    Assay 99.994%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H272-H315-H319
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit