Zirconium hydride, 99.7% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 89484-30

Quantity 250g

 View more versions of this product

Catalog No. AA8948430

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7704-99-6
    Physical Form -325 Mesh Powder
    Assay Percent Range 99.7% (metals basis)
    Molecular Formula ZrH2
    Formula Weight 93.24
    MDL Number MFCD00148878
    Chemical Name or Material Zirconium hydride

    Specifications

    Formula Weight 93.24
    Quantity 250g
    EINECS Number 231-727-3
    Physical Form -325 Mesh Powder
    Density 5.47g/mL
    Assay Percent Range 99.7% (metals basis)
    Molecular Formula ZrH2
    MDL Number MFCD00148878
    CAS 7704-99-6
    Chemical Name or Material Zirconium hydride

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit