Alfa Aesar™ Zirconium(IV) 1,1,1-trifluoro-2,4-pentanedionate, 98+%

Manufacturer: Alfa Aesar™ 1927606

Quantity 5g

 View more versions of this product

Catalog No. AA1927606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17499-68-2
    Formula Weight 703.46
    MDL Number MFCD00015317
    Synonym Zirconium(IV) trifluoroacetylacetonate
    Chemical Name or Material Zirconium(IV) 1,1,1-trifluoro-2,4-pentanedionate

    Specifications

    Formula Weight 703.46
    CAS 17499-68-2
    Synonym Zirconium(IV) trifluoroacetylacetonate
    Solubility Information Soluble in toluene; Slightly soluble in THF and methylene chloride; Insoluble in water
    Physical Form Crystalline
    MDL Number MFCD00015317
    Quantity 5g
    Boiling Point 235°C (dec.)
    Chemical Name or Material Zirconium(IV) 1,1,1-trifluoro-2,4-pentanedionate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit