Learn More
Zirconium(IV) chloride, Reactor Grade, 99.5+% (metals basis), Thermo Scientific Chemicals
Description
Zirconium tetrachloride is commonly utilized as a Lewis acid catalyst in many organic synthesis reactions. It is an important source for organometallic complexes of zirconium. It is practiced in many areas like nuclear industry, metal industry, photography, glass, and ceramic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 331° (sublimation) |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | >99.5% (metals basis) |
Solubility Information | Soluble in alcohol,ether,hydrochloric acid. |
Formula Weight | 233.03 |
Grade | Reactor |
Sensitivity | Moisture sensitive |
Density | 2.803 |
Chemical Name or Material | Zirconium(IV) chloride |
Safety and Handling
Hazard Category | H290-H302-H314 |
Precautionary Statement | P234-P264b-P270-P280-P284-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P390-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 233-058-2 |
RTECSNumber | ZH7175000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.