Zirconium(IV) iodide, 99%, Alfa Aesar™

Light, Air and Moisture Sensitive

Manufacturer: Alfa Aesar 1740006

Quantity 5g

 View more versions of this product

Catalog No. AA1740006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13986-26-0
    Physical Form Powder
    Assay Percent Range 99%
    Molecular Formula I4Zr
    Formula Weight 598.84
    MDL Number MFCD00049641
    Synonym Zirconium tetraiodide

    Specifications

    Formula Weight 598.84
    Quantity 5g
    Color Yellow
    Physical Form Powder
    Assay Percent Range 99%
    Molecular Formula I4Zr
    MDL Number MFCD00049641
    CAS 13986-26-0
    Synonym Zirconium tetraiodide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit