Alfa Aesar™ Zirconium(IV) isopropoxide isopropanol complex

Manufacturer: Alfa Aesar™ 8611818

Quantity 50g

 View more versions of this product

Catalog No. AA8611818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14717-56-7
    Molecular Formula Zr[OCH(CH3)2]4·(CH3)2CHOH
    Formula Weight 387.67
    MDL Number MFCD00075157
    UN Number UN3181
    Chemical Name or Material Zirconium (IV) isopropoxide isopropanol complex

    Specifications

    Formula Weight 387.67
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Moisture Sensitive
    UN Number UN3181
    Quantity 50g
    Solubility Information Soluble in hot Isopropanol, warm hexane and toluene; Tends to precipitate over time
    Physical Form Crystalline
    MDL Number MFCD00075157
    Health Hazard 1 Warning
    Molecular Formula Zr[OCH(CH3)2]4·(CH3)2CHOH
    CAS 14717-56-7
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Chemical Name or Material Zirconium (IV) isopropoxide isopropanol complex

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit