Alfa Aesar™ Zirconium(IV) oxide 2-ethylhexanoate, in mineral spirits (»6% Zr)

Manufacturer: Alfa Aesar™ 3954322

Quantity 100g

 View more versions of this product

Catalog No. AA3954322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22464-99-9
    MDL Number MFCD00072684
    UN Number UN1268
    Synonym Zirconyl 2-ethylhexanoate
    Chemical Name or Material Zirconium(IV) oxide 2-ethylhexanoate, In mineral spirits

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00072684
    CAS 22464-99-9
    UN Number UN1268
    Synonym Zirconyl 2-ethylhexanoate
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Quantity 100g
    Flash Point 40° (104°F)
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
    Concentration ≈6% Zr
    Chemical Name or Material Zirconium(IV) oxide 2-ethylhexanoate, In mineral spirits

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit