Zirconium(IV) oxide, 20% in H2O, colloidal dispersion, Alfa Aesar™

Manufacturer: Alfa Aesar 1273236

Quantity 500g

 View more versions of this product

Catalog No. AA1273236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-23-4
    Physical Form Liquid
    Assay Percent Range 20% in H2O
    Molecular Formula ZrO2
    Formula Weight 123.22
    MDL Number MFCD00011310
    Chemical Name or Material colloidal dispersion

    Specifications

    Formula Weight 123.22
    Quantity 500g
    Color Off-white
    Physical Form Liquid
    Odor Recognizable
    Assay Percent Range 20% in H2O
    Molecular Formula ZrO2
    MDL Number MFCD00011310
    CAS 1314-23-4
    Chemical Name or Material colloidal dispersion

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit