Alfa Aesar™ Zirconium(IV) oxide, 99.5% (metals basis excluding Hf), Hf <100ppm

Manufacturer: Alfa Aesar™ 4014036

Quantity 500g

 View more versions of this product

Catalog No. AA4014036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-23-4
    Formula Weight 123.22
    MDL Number MFCD00011310
    Merck Index 14,10180
    Chemical Name or Material Zirconium(IV) oxide

    Specifications

    Formula Weight 123.22
    Merck Index 14,10180
    Color White
    Density 5.89g/mL
    RTECS Number ZH8800000
    Physical Form Powder
    Boiling Point ≈5000°C
    Melting Point ≈2700°C
    Quantity 500g
    TSCA TSCA
    Odor Odorless
    Particle Size 1 Micron APS
    MDL Number MFCD00011310
    CAS 1314-23-4
    Chemical Name or Material Zirconium(IV) oxide
    Assay 99.5%, (metals basis excluding Hf), Hf <100ppm

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit