Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium(IV) oxide, 99.5% (metals basis excluding Hf), Hf <100ppm

Manufacturer:  Alfa Aesar™ 4014022

Quantity 100g

 View more versions of this product

Catalog No. AA4014022


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1314-23-4
MFCD00011310
Zirconium(IV) oxide
123.22
14,10180

Specifications

1314-23-4
123.22
14,10180
Odorless
≈5000°C
≈2700°C
Zirconium(IV) oxide
Powder
MFCD00011310
5.89g/mL
1 Micron APS
White
100g
99.5%, (metals basis excluding Hf), Hf <100ppm

Safety and Handling

SDS & Safety Documents

RTECSNumber : ZH8800000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit