Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium(IV) oxide, Aerosol Refractory Paint, 99% (including 4% CaO)

Manufacturer:  Alfa Aesar™ 40480CLC

Quantity 13 oz.

 View more versions of this product

Catalog No. AA40480CLC

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1314-23-4
UN1950
Zirconium(IV) oxide, Aerosol Refractory Paint
MFCD00011310
14,10180

Specifications

1314-23-4
UN1950
Aerosol Spray Can
13 oz.
99%, including 4% CaO
MFCD00011310
14,10180
Cream
Zirconium(IV) oxide, Aerosol Refractory Paint

Safety and Handling

SDS & Safety Documents

GHS H Statement
H222-H319
Extremely flammable aerosol.
Causes serious eye irritation.

GHS P Statement
P210-P251-P211-P280-P305+P351+P338-P410+P412
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Pressurized container: Do not pierce or burn, even after use.
Do not spray on an open flame or other ignition source.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Protect from sunlight. Do no expose to temperatures exceeding 50 celcius degress.

Danger

DOTInformation : Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit