Alfa Aesar™ Zirconium(IV) oxide, Aerosol Refractory Paint, 99% (including 4% CaO)

Manufacturer: Alfa Aesar™ 404800CL

Quantity 5 x 13 oz.

 View more versions of this product

Catalog No. AA404800CL

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-23-4
    MDL Number MFCD00011310
    UN Number UN1950
    Merck Index 14,10180
    Chemical Name or Material Zirconium(IV) oxide, Aerosol Refractory Paint

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P251-P211-P280-P305+P351+P338-P410+P412
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Pressurized container: Do not pierce or burn, even after use.
    Do not spray on an open flame or other ignition source.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Protect from sunlight. Do no expose to temperatures exceeding 50 celcius degress.
    TSCA TSCA
    MDL Number MFCD00011310
    CAS 1314-23-4
    UN Number UN1950
    Merck Index 14,10180
    Color Cream
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS
    Packaging Aerosol Spray Can
    Quantity 5 x 13 oz.
    Health Hazard 2 GHS H Statement
    H222-H319
    Extremely flammable aerosol.
    Causes serious eye irritation.
    Chemical Name or Material Zirconium(IV) oxide, Aerosol Refractory Paint
    Assay 99%, including 4% CaO

    GHS H Statement
    H222-H319
    Extremely flammable aerosol.
    Causes serious eye irritation.

    GHS P Statement
    P210-P251-P211-P280-P305+P351+P338-P410+P412
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Pressurized container: Do not pierce or burn, even after use.
    Do not spray on an open flame or other ignition source.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Protect from sunlight. Do no expose to temperatures exceeding 50 celcius degress.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit