Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium(IV) oxide, calcia stabilized, 99.4% (metals basis excluding Hf)

Click to view available options
Quantity:
50g
250g
1kg
1314-23-4
O2Zr
123.222
MFCD00011310
MCMNRKCIXSYSNV-UHFFFAOYSA-N
zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler
62395
dioxozirconium
O=[Zr]=O
1314-23-4
O2Zr
123.222
MFCD00011310
MCMNRKCIXSYSNV-UHFFFAOYSA-N
zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler
62395
dioxozirconium
O=[Zr]=O
Merck Index 14,10180
Formula Weight 123.22
Physical Form -100+325 Mesh Powder
Percent Purity 99.4%
Concentration ZrO2, 4% CaO
Mesh Size −100+325 Mesh
Odor Odorless
Boiling Point ~5000°C
Melting Point ~2700°C
Quantity 250g
Chemical Name or Material Zirconium(IV) oxide, Calcia stabilized
Assay (metals basis excluding Hf)
Show More Show Less

EINECSNumber : 215-227-2

RTECSNumber : ZH8800000

TSCA : Yes

Recommended Storage : Ambient temperatures