Zirconium(IV) oxide, cement, Alfa Aesar™

One component adhesive. Bonds to Zirconium oxide and Silicon. Exhibit good dielectric properties

Manufacturer: Alfa Aesar 42493A7

Quantity 5kg

 View more versions of this product

Catalog No. AA42493A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-23-4
    MDL Number MFCD01868884
    Chemical Name or Material Zirconium(IV) oxide

    Specifications

    Quantity 5kg
    Color Light brown
    EINECS Number 215-227-2
    Odor Odorless
    Appearance Light brown liquid/paste
    MDL Number MFCD01868884
    CAS 1314-23-4
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Zirconium(IV) oxide

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit