Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium(IV) oxide, Puratronic™, 99.978% (metals basis)

Manufacturer:  Alfa Aesar™ 1114136

Quantity 500g

 View more versions of this product

Catalog No. AA1114136


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1314-23-4
123.22
14,10180
Puratronic™
ZrO2
MFCD00011310
Zirconium(IV) oxide

Specifications

1314-23-4
ZrO2
MFCD00011310
5.89g/mL
≈5000°C
≈2700°C
Zirconium(IV) oxide
Puratronic™
Powder
123.22
14,10180
Odorless
White
500g
99.978%, (metals basis)

Safety and Handling

SDS & Safety Documents

RTECSNumber : ZH8800000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit