Alfa Aesar™ Zirconium(IV) oxide, titania stablized

Manufacturer: Alfa Aesar™ 4512622

Quantity 100g

 View more versions of this product

Catalog No. AA4512622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-23-4
    MDL Number MFCD00011310
    Merck Index 14,10180
    Chemical Name or Material Zirconium (IV) oxide, Titania stablized

    Specifications

    Odor Odorless
    Physical Form Tablets
    CAS 1314-23-4
    Color Black
    Weight ≈1g Tablets
    Avg. Mol. Wt. or Mol. Wt. Range ZrO2:TiO2, 90:10 wt%
    Density 5.4g/mL
    TSCA TSCA
    Merck Index 14,10180
    Melting Point ≈2500°C
    Quantity 100g
    RTECS Number ZH8800000
    MDL Number MFCD00011310
    Chemical Name or Material Zirconium (IV) oxide, Titania stablized

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit