Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) Oxide, Yttria Stabilized, 99% (Metal Basis excluding Hf), Alfa Aesar™

Click to view available options
Quantity:
100g
500g
1314-23-4
O2Zr
123.222
MFCD00145546
MCMNRKCIXSYSNV-UHFFFAOYSA-N
zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler
62395
dioxozirconium
O=[Zr]=O
1314-23-4
O2Zr
123.222
MFCD00145546
MCMNRKCIXSYSNV-UHFFFAOYSA-N
zirconia,zirconium oxide,zirconium oxide zro2,rhuligel,zirconium iv oxide,zirconium white,zirconic anhydride,pigment white 12,zirox zt 35,pcs filler
62395
dioxozirconium
O=[Zr]=O

Stabilized with 1.3% yttrium

Assay Percent Range 99%, metal basis excluding Hf
Solubility Information Insoluble in water. Soluble in hot and concentrated hydrofluoric acid and sulfuric acid.
Physical Form -325 Mesh Powder
Boiling Point ca 5000°C
Melting Point ∽2700°C
Quantity 100g
Chemical Name or Material Zirconium(IV) Oxide

EINECSNumber : 215-227-2

RTECSNumber : ZH8800000

TSCA : Yes

Recommended Storage : Ambient temperatures