Learn More
Zirconium(IV) tert-butoxide, 99.99% (metals basis), Thermo Scientific Chemicals
Description
It finds it application as a reagent for the preparation of heterometallic tin(II) or lead(II) zirconium alkoxides. It is used as a catalyst in cyanation process. Zirconium tert-butoxide (ZTB) precursor is used to deposit thin films of Zirconia and other zirconium containing films by atomic layer deposition and chemical vapor deposition methods. Zirconium oxide thin films can also be grown at low temperatures, ranging from 150°C to 300°C, by the presence of moisture along with ZTB or by UV-enhanced atomic layer deposition (UV-ALD) process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 0.985 g/mL |
Boiling Point | 90°C (5 mmHg) |
Flash Point | 85°C (185°F) |
Linear Formula | Zr[OC(CH3)3]4 |
Quantity | 2 g |
Beilstein | 3681870 |
Sensitivity | Light and moisture sensitive |
Solubility Information | Decomposes in water. |
Formula Weight | 383.68 |
Show More |
Safety and Handling
Hazard Category | H226 |
Hazard Statement | GHS H Statement H315-H319-H227 Causes skin irritation. Causes serious eye irritation. Combustible liquid. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Hazard Class: 3; Packaging Group: II |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.