Alfa Aesar™ Zirconium Nickel powder, -325 mesh

Manufacturer: Alfa Aesar™ 8840636

Quantity 500g

 View more versions of this product

Catalog No. AA8840636

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00192582
    UN Number UN3089
    Chemical Name or Material Zirconium Nickel powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −325 Mesh
    Health Hazard 1 Danger
    Quantity 500g
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00192582
    UN Number UN3089
    Health Hazard 2 GHS H Statement
    H228-H351-H317
    Flammable solid.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Sensitivity Air Sensitive
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    Avg. Mol. Wt. or Mol. Wt. Range Zr:Ni; 70:30 wt%
    Color Dark gray
    Chemical Name or Material Zirconium Nickel powder

    GHS H Statement
    H228-H351-H317
    Flammable solid.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit