Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium Nickel powder, -325 mesh

$150.60 - $573.50

Specifications

Chemical Name or Material Zirconium Nickel powder
MDL Number MFCD00192582
UN Number UN3089
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA8840622 View Documents Alfa Aesar™
8840622
100g Each for $150.60
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA8840636 View Documents Alfa Aesar™
8840636
500g Each for $573.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Zirconium Nickel powder
MFCD00192582
UN3089
Safety and Handling

Safety and Handling

GHS H Statement
H228-H351-H317
Flammable solid.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P210-P240-P241-P261-P281-P285-P304+P341-P308+P313-P342+P311-P370+P378q-P501c

H228-H334-H351

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit