Learn More
Alfa Aesar™ Zirconium Nickel powder, -325 mesh
Manufacturer:  Alfa Aesar™ 8840614
Quantity 25g
 View more versions of this product
Catalog No. AA8840614
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
MFCD00192582
UN3089
Zirconium Nickel powder
Specifications
Zr:Ni; 70:30 wt%
Powder
UN3089
Air Sensitive
Zirconium Nickel powder
25g
MFCD00192582
−325 Mesh
Dark gray
Safety and Handling
SDS & Safety Documents
GHS H Statement
H228-H351-H317
Flammable solid.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
TSCA : TSCA