Zirconium Oxide, Catalyst Support, Tungstated, Alfa Aesar™

Manufacturer: Alfa Aesar 45597A4

Quantity 2.5kg

 View more versions of this product

Catalog No. AA45597A4

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00168096
    Chemical Name or Material Zirconium Oxide

    Specifications

    Quantity 2.5kg
    Particle Size 3mm
    MDL Number MFCD00168096
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Zirconium Oxide

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit