Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium oxide, polymeric precursor

Preparation of zirconium oxide films, solid electrolyte

Manufacturer:  Alfa Aesar™ 4276614

Quantity 25g

 View more versions of this product

Catalog No. AA4276614


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00011310
UN1993
Zirconium oxide, Polymeric precursor

Specifications

Viscous Liquid
MFCD00011310
Alcohol-like
Moisture Sensitive
Zirconium oxide, Polymeric precursor
Yellow to brown
25g
UN1993
Soluble in alcohols

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H315-H319-H335-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit