Zirconium oxide, Yttria stabilized, Grinding Pieces, 5mm diameter, Alfa Aesar™

Specifications: Yttrium oxide (mole) 3%, Hardness 12, Total impuroty level <0.1

Manufacturer: Alfa Aesar 4328530

Quantity 250g

 View more versions of this product

Catalog No. AA4328530

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00145546
    Chemical Name or Material Zirconium oxide

    Specifications

    Quantity 250g
    Color Off-white
    Odor Odorless
    Appearance Off-white chunks
    MDL Number MFCD00145546
    Chemical Name or Material Zirconium oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit