Learn More
Zirconium oxide Yttrium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 038509KT
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium Oxide Yttrium Oxide Substrate | |
Plate | |
ZrO2 + 9.5% Y2O3 | |
Odorless |
Polished One Side | |
1 Ea. | |
MFCD01868885 |
Safety and Handling
EINECSNumber : 215-227-2
TSCA : Yes
Recommended Storage : Ambient temperatures