Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium plate, 6.4mm (0.25 in.) thick, 25x100mm (0.98x3.9 in.), 99.2% (metals basis excluding Hf)

Click to view available options
Quantity:
1 pc.
5 pc.
7440-67-7
Zr
91.224
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil
23995
CHEBI:33342
zirconium
[Zr]
7440-67-7
Zr
91.224
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil
23995
CHEBI:33342
zirconium
[Zr]
Quantity 5 pc.
Weight ≈104g/25x100mm
Odor Odorless
Physical Form Plate
Percent Purity 99.2%
Chemical Name or Material Zirconium plate, 6.4mm (0.25 in.) thick, 25x100mm (0.98x3.9 in.), Temper: annealed
Assay (metals basis excluding Hf)

EINECSNumber : 231-176-9

TSCA : Yes

Recommended Storage : Ambient temperatures