Zirconium powder, Alfa Aesar™

Manufacturer: Alfa Aesar 0041888

Quantity 2.5g

 View more versions of this product

Catalog No. AA0041888

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-67-7
    Physical Form Powder
    MDL Number MFCD00011303
    Chemical Name or Material Zirconium

    Specifications

    Quantity 2.5g
    Color Dark gray
    DOT Information DOT Class: 4.1, Packaging Group: II
    EINECS Number 231-176-9
    Physical Form Powder
    MDL Number MFCD00011303
    CAS 7440-67-7
    Odor Odorless
    Health Hazard 1 H228: Flammable solid.
    Appearance Dark Gray powder
    Vapor Pressure 23 hPa (17mmHg)
    Chemical Name or Material Zirconium

    H228: Flammable solid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit