Zirconium Sponge, 25.4mm (1.0 in.) and down, 99.2% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1419136

Quantity 500g

 View more versions of this product

Catalog No. AA1419136

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7440-67-7
    Physical Form Sponge, 25.4mm (1.0 in.) and Down
    Assay Percent Range 99.2% (Metals basis)
    MDL Number MFCD00011303
    Chemical Name or Material Zirconium

    Specifications

    Physical Form Sponge, 25.4mm (1.0 in.) and Down
    Quantity 500g
    Color Gray
    DOT Information DOT Class: 4.2, Packaging Group: III
    EINECS Number 231-176-9
    Assay Percent Range 99.2% (Metals basis)
    MDL Number MFCD00011303
    CAS 7440-67-7
    Odor Odorless
    Health Hazard 1 H260-H252: In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities; may catch fire.
    Appearance Gray sponge
    Chemical Name or Material Zirconium

    H260-H252: In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities; may catch fire.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit