Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium Standard, 1mL = 1mg Zr, 1000ppm Zr, Ricca Chemical
SDP

Catalog Number p-3252649
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Poly natural
13826-66-9, 7697-37-2, 7732-18-5
H2N2O7Zr
233.247
UJVRJBAUJYZFIX-UHFFFAOYSA-N
83761
nitric acid;oxozirconium
[N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
13826-66-9, 7697-37-2, 7732-18-5
H2N2O7Zr
233.247
UJVRJBAUJYZFIX-UHFFFAOYSA-N
83761
nitric acid;oxozirconium
[N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
Packaging Poly natural
Quantity 120 mL
Color Colorless
Concentration or Composition Notes 1,000ppm Zr
Equivalence 1 mL = 1 mg Zr
Physical Form Liquid
Chemical Name or Material Zirconium Standard
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit