Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
7440-67-7,7664-39-3,7732-18-5
Zr
91.224
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
CHEBI:33342
zirconium
[Zr]
7440-67-7,7664-39-3,7732-18-5
Zr
91.224
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
CHEBI:33342
zirconium
[Zr]
for atomic adsorption spectroscopy
Concentration 990 to 1010μg/mL of Zr(4+) in 5% HF
Physical Form Liquid
Grade AAS
Color Colorless
Quantity 500mL
Chemical Name or Material Zirconium standard solution, 1 mg/ml Zr in 5% HF

GHS H Statement:
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger