Alfa Aesar™ Zirconium telluride, 99.5% (metals basis excluding Hf), Hf 2-4%

Manufacturer: Alfa Aesar™ 3632114

Quantity 25g

 View more versions of this product

Catalog No. AA3632114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32321-65-6
    Formula Weight 346.42
    MDL Number MFCD00145549
    Chemical Name or Material Zirconium telluride

    Specifications

    Formula Weight 346.42
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −325 Mesh
    CAS 32321-65-6
    Color Gray
    Quantity 25g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Density 6.42g/mL
    TSCA TSCA
    MDL Number MFCD00145549
    Physical Form Powder
    Chemical Name or Material Zirconium telluride
    Assay 99.5%, (metals basis excluding Hf), Hf 2 to 4%

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit