Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate), 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
18865-74-2
C44H80O8Zr
828.34
MFCD00145380
ANBXKEDJQLYCLX-UHFFFAOYSA-N
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium,zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate,tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv,zr tmhd 4,zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate,zirconium tetrakis dipivaloylmethanate,zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate,4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one
50919870
5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Zr]
18865-74-2
C44H80O8Zr
828.34
MFCD00145380
ANBXKEDJQLYCLX-UHFFFAOYSA-N
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium,zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate,tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv,zr tmhd 4,zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate,zirconium tetrakis dipivaloylmethanate,zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate,4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one
50919870
5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Zr]
Elemental Analysis 98.5% min. ( ex Zr )
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Assay Percent Range 99%
Formula Weight 824.3
Physical Form Crystalline Powder
Chemical Name or Material Zirconium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate)

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.